Assay Details
| Gene Symbol: serpinh1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001309823.1 | NP_001296752.1 | 2 - 3 | 761 | 104 | |
| NM_131204.2 | NP_571279.2 | 3 - 4 | 809 | 104 | ||
| GenBank mRNA | U31079.1 | - | 3 - 4 | 749 | 104 | |
More Information
Set Membership: |
|

serpinh1b
Genomic Map