Assay Details
| Gene Symbol: igf2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131433.1 | NP_571508.1 | 1 - 2 | 190 | 107 | |
| GenBank mRNA | AF194333.1 | - | 1 - 2 | 190 | 107 | |
| BC085623.1 | - | 1 - 2 | 174 | 107 | ||
More Information
Set Membership: |
|

igf2a
Genomic Map