Assay Details
| Gene Symbol: vsx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131462.2 | NP_571537.2 | 1 - 2 | 490 | 90 | |
| XM_005158805.4 | XP_005158862.1 | 1 - 2 | 735 | 90 | ||
| GenBank mRNA | BC055588.1 | - | 1 - 2 | 490 | 90 | |
| BC067664.1 | - | 1 - 2 | 452 | 90 | ||
| BC164202.1 | - | 1 - 2 | 452 | 90 | ||
| U62898.1 | - | 1 - 2 | 484 | 90 | ||
More Information
Set Membership: |
|

vsx2
Genomic Map