Assay Details
| Gene Symbol: smad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131571.2 | NP_571646.1 | 2 - 3 | 596 | 105 | |
| GenBank mRNA | AF229023.1 | - | 2 - 3 | 354 | 105 | |
| AY134492.1 | - | 2 - 3 | 485 | 105 | ||
| BC129150.1 | - | 2 - 3 | 596 | 105 | ||
More Information
Set Membership: |
|

smad3a
Genomic Map