Assay Details
| Gene Symbol: smad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131571.2 | NP_571646.1 | 6 - 7 | 1076 | 95 | |
| GenBank mRNA | AY134492.1 | - | 6 - 7 | 965 | 95 | |
| BC129150.1 | - | 6 - 7 | 1076 | 95 | ||
More Information
Set Membership: |
|

smad3a
Genomic Map