Assay Details
| Gene Symbol: smad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131571.2 | NP_571646.1 | 7 - 8 | 1212 | 100 | |
| GenBank mRNA | AY134492.1 | - | 7 - 8 | 1101 | 100 | |
| BC129150.1 | - | 7 - 8 | 1212 | 100 | ||
More Information
Set Membership: |
|

smad3a
Genomic Map