Assay Details
| Gene Symbol: cd74a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131590.1 | NP_571665.1 | 5 - 6 | 565 | 66 | |
| GenBank mRNA | AF114830.1 | - | 5 - 6 | 562 | 66 | |
| AF148214.1 | - | 5 - 6 | 565 | 66 | ||
| BC065433.1 | - | 5 - 6 | 555 | 66 | ||
More Information
Set Membership: |
|

cd74a
Genomic Map