Assay Details
| Gene Symbol: ctbp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195491.1 | NP_001182420.1 | 5 - 6 | 2795 | 117 | |
| NM_131715.1 | NP_571790.1 | 4 - 5 | 605 | 117 | ||
| GenBank mRNA | AB032416.1 | - | 4 - 5 | 605 | 117 | |
| AY878349.1 | - | 5 - 6 | 2795 | 117 | ||
| AY878350.1 | - | 4 - 5 | 1364 | 117 | ||
| BC045361.1 | - | 5 - 6 | 784 | 117 | ||
| BC056327.1 | - | 5 - 6 | 800 | 117 | ||
| BC066374.1 | - | 5 - 6 | 853 | 117 | ||
| BC066380.1 | - | 5 - 6 | 946 | 117 | ||
| BC155842.1 | - | 4 - 5 | 1740 | 117 | ||
| BC164785.1 | - | 4 - 5 | 600 | 117 | ||
More Information
Set Membership: |
|

ctbp2a
Genomic Map