Assay Details
| Gene Symbol: ctbp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131715.1 | NP_571790.1 | 8 - 9 | 1196 | 75 | |
| GenBank mRNA | AB032416.1 | - | 8 - 9 | 1196 | 75 | |
| BC155842.1 | - | 8 - 9 | 2331 | 75 | ||
More Information
Set Membership: |
|

ctbp2a
Genomic Map