Assay Details
| Gene Symbol: cldn15la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131771.2 | NP_571846.2 | 3 - 4 | 557 | 83 | |
| GenBank mRNA | AF359427.1 | - | 3 - 4 | 518 | 83 | |
| BC093140.1 | - | 3 - 4 | 508 | 83 | ||
| EH495796.1 | - | 3 - 4 | 588 | 83 | ||
| EH507578.1 | - | 3 - 4 | 566 | 83 | ||
| EH533727.1 | - | 2 - 3 | 380 | 83 | ||
More Information
Set Membership: |
|

cldn15la
Genomic Map