Assay Details
| Gene Symbol: cldn15la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131771.2 | NP_571846.2 | 4 - 5 | 669 | 95 | |
| GenBank mRNA | AF359427.1 | - | 4 - 5 | 630 | 95 | |
| BC093140.1 | - | 4 - 5 | 620 | 95 | ||
| EH495796.1 | - | 4 - 5 | 700 | 95 | ||
| EH507578.1 | - | 4 - 5 | 678 | 95 | ||
| EH533727.1 | - | 1 - 2 | 268 | 95 | ||
More Information
Set Membership: |
|

cldn15la
Genomic Map