Assay Details
| Gene Symbol: gng3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_131841.2 | NP_571916.1 | 2 - 3 | 178 | 77 | |
| GenBank mRNA | AF364083.1 | - | 2 - 3 | 237 | 77 | |
| BC076314.1 | - | 2 - 3 | 178 | 77 | ||
| BC164230.1 | - | 131 | 77 | |||
More Information
Set Membership: |
|

gng3
Genomic Map