Assay Details
| Gene Symbol: adcyap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001328496.1 | NP_001315425.1 | 4 - 5 | 806 | 78 | |
| NM_152885.2 | NP_690841.1 | 4 - 5 | 806 | 78 | ||
| GenBank mRNA | BC083483.1 | - | 4 - 5 | 380 | 78 | |
| BC164998.1 | - | 3 - 4 | 367 | 78 | ||
More Information
Set Membership: |
|

adcyap1a
Genomic Map