Assay Details
| Gene Symbol: esr2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174862.3 | NP_777287.2 | 8 - 9 | 1770 | 96 | |
| XM_005156971.4 | XP_005157028.1 | 9 - 10 | 2103 | 96 | ||
| XM_005156972.4 | XP_005157029.1 | 10 - 11 | 2277 | 96 | ||
| XM_005156973.4 | XP_005157030.1 | 9 - 10 | 1790 | 96 | ||
| GenBank mRNA | AF349413.3 | - | 9 - 10 | 1789 | 96 | |
| AJ275911.3 | - | 9 - 10 | 1834 | 96 | ||
| AJ414566.1 | - | 9 - 10 | 1790 | 96 | ||
| BC086848.1 | - | 8 - 9 | 1770 | 96 | ||
| BC164792.1 | - | 8 - 9 | 1745 | 96 | ||
More Information
Set Membership: |
|

esr2b
Genomic Map