Assay Details
| Gene Symbol: tfap2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001319158.1 | NP_001306087.1 | 6 - 7 | 1048 | 92 | |
| NM_001319159.2 | NP_001306088.1 | 6 - 7 | 1452 | 92 | ||
| NM_176859.4 | NP_789829.2 | 6 - 7 | 1098 | 92 | ||
| XM_005162558.4 | XP_005162615.1 | 6 - 7 | 1270 | 92 | ||
| GenBank mRNA | AF457191.1 | - | 6 - 7 | 1076 | 92 | |
| AF457192.1 | - | 6 - 7 | 1056 | 92 | ||
| AY166856.1 | - | 6 - 7 | 1370 | 92 | ||
| BC060900.1 | - | 6 - 7 | 1267 | 92 | ||
| BC066582.1 | - | 6 - 7 | 1354 | 92 | ||
| BC090506.1 | - | 3 - 4 | 1189 | 92 | ||
| EH590405.1 | - | 1 - 2 | 599 | 92 | ||
More Information
Set Membership: |
|

tfap2a
Genomic Map