Assay Details
| Gene Symbol: sim1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178222.3 | NP_835740.2 | 8 - 9 | 1254 | 89 | |
| GenBank mRNA | AY028626.1 | - | 8 - 9 | 1263 | 89 | |
More Information
Set Membership: |
|

sim1a
Genomic Map