Assay Details
| Gene Symbol: otx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181331.3 | NP_851848.2 | 2 - 3 | 445 | 75 | |
| GenBank mRNA | AY036005.1 | - | 2 - 3 | 464 | 75 | |
| BC066464.1 | - | 2 - 3 | 445 | 75 | ||
More Information
Set Membership: |
|

otx5
Genomic Map