Assay Details
| Gene Symbol: tcerg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198357.3 | NP_938171.2 | 16 - 17 | 2445 | 62 | |
| GenBank mRNA | BC056813.1 | - | 16 - 17 | 2461 | 62 | |
| CN506442.1 | - | 2 - 3 | 288 | 62 | ||
More Information
Set Membership: |
|

tcerg1a
Genomic Map