Assay Details
| Gene Symbol: tcerg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198357.3 | NP_938171.2 | 1 - 2 | 54 | 93 | |
| GenBank mRNA | BC056813.1 | - | 1 - 2 | 70 | 93 | |
| Gene Symbol: LOC103910002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009298386.3 | XP_009296661.2 | 1 - 2 | 83 | 93 | |
| XM_021471000.1 | XP_021326675.1 | 1 - 2 | 83 | 93 | ||
More Information
Set Membership: |
|

tcerg1a
Genomic Map