Assay Details
| Gene Symbol: tcerg1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_198357.3 | NP_938171.2 | 10 - 11 | 1606 | 57 | |
| GenBank mRNA | BC056813.1 | - | 10 - 11 | 1622 | 57 | |
More Information
Set Membership: |
|

tcerg1a
Genomic Map