Assay Details
| Gene Symbol: col1a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199214.1 | NP_954684.1 | 29 - 30 | 1942 | 88 | |
| GenBank mRNA | AY380817.1 | - | 29 - 30 | 1942 | 88 | |
| BC063249.1 | - | 29 - 30 | 2037 | 88 | ||
| BC161663.1 | - | 29 - 30 | 2054 | 88 | ||
More Information
Set Membership: |
|

col1a1a
Genomic Map