Assay Details
| Gene Symbol: col1a1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199214.1 | NP_954684.1 | 7 - 8 | 538 | 105 | |
| GenBank mRNA | AY380817.1 | - | 7 - 8 | 538 | 105 | |
| BC063249.1 | - | 7 - 8 | 633 | 105 | ||
| BC161663.1 | - | 7 - 8 | 650 | 105 | ||
More Information
Set Membership: |
|

col1a1a
Genomic Map