Assay Details
| Gene Symbol: lgi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199627.2 | NP_955921.2 | 1 - 2 | 339 | 72 | |
| GenBank mRNA | AY309261.1 | - | 1 - 2 | 281 | 72 | |
| BC056753.1 | - | 1 - 2 | 322 | 72 | ||
| BC065665.1 | - | 1 - 2 | 303 | 72 | ||
| CU468780.1 | - | 1 - 2 | 171 | 72 | ||
| CU638810.1 | - | 1 - 2 | 184 | 72 | ||
More Information
Set Membership: |
|

lgi1a
Genomic Map