Assay Details
| Gene Symbol: lgi1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199627.2 | NP_955921.2 | 6 - 7 | 797 | 103 | |
| GenBank mRNA | BC056753.1 | - | 6 - 7 | 780 | 103 | |
| BC065665.1 | - | 6 - 7 | 761 | 103 | ||
| CU638810.1 | - | 6 - 7 | 642 | 103 | ||
More Information
Set Membership: |
|

lgi1a
Genomic Map