Assay Details
| Gene Symbol: rpl10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199636.1 | NP_955930.1 | 4 - 5 | 333 | 84 | |
| XM_021478186.1 | XP_021333861.1 | 4 - 5 | 534 | 84 | ||
| GenBank mRNA | BC059454.1 | - | 4 - 5 | 333 | 84 | |
| BC071510.1 | - | 4 - 5 | 350 | 84 | ||
More Information
Set Membership: |
|

rpl10a
Genomic Map