Assay Details
| Gene Symbol: srsf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199651.2 | NP_955945.2 | 1 - 2 | 544 | 161 | |
| NR_132430.1 | 1 - 2 | 544 | 161 | |||
| GenBank mRNA | BC045480.1 | - | 1 - 2 | 500 | 155 | |
| BC065971.1 | - | 1 - 2 | 486 | 161 | ||
| EH606115.1 | - | 3 - 4 | 652 | 158 | ||
More Information
Set Membership: |
|

srsf2b
Genomic Map