Assay Details
| Gene Symbol: srsf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199651.2 | NP_955945.2 | 2 - 3 | 847 | 66 | |
| NR_132430.1 | 2 - 3 | 847 | 66 | |||
| GenBank mRNA | BC045480.1 | - | 2 - 3 | 803 | 66 | |
| BC065971.1 | - | 2 - 3 | 789 | 66 | ||
| EH606115.1 | - | 2 - 3 | 349 | 66 | ||
More Information
Set Membership: |
|

srsf2b
Genomic Map