Assay Details
| Gene Symbol: atp5f1c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200041.2 | NP_956335.2 | 8 - 9 | 927 | 83 | |
| GenBank mRNA | BC060917.1 | - | 8 - 9 | 897 | 83 | |
| EH461834.1 | - | 1 - 2 | 224 | 83 | ||
| EH528823.1 | - | 1 - 2 | 226 | 83 | ||
More Information
Set Membership: |
|

atp5f1c
Genomic Map