Assay Details
| Gene Symbol: mars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200076.1 | NP_956370.1 | 11 - 12 | 1546 | 97 | |
| GenBank mRNA | AY423024.1 | - | 11 - 12 | 1546 | 97 | |
More Information
Set Membership: |
|

mars1
Genomic Map