Assay Details
| Gene Symbol: mars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200076.1 | NP_956370.1 | 17 - 18 | 2382 | 82 | |
| GenBank mRNA | AY423024.1 | - | 17 - 18 | 2382 | 82 | |
| AY648846.1 | - | 17 - 18 | 2382 | 82 | ||
| BC046899.1 | - | 17 - 18 | 2382 | 82 | ||
| BC057463.1 | - | 17 - 18 | 2363 | 82 | ||
More Information
Set Membership: |
|

mars1
Genomic Map