Assay Details
| Gene Symbol: mars1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200076.1 | NP_956370.1 | 6 - 7 | 839 | 84 | |
| GenBank mRNA | AY423024.1 | - | 6 - 7 | 839 | 84 | |
| AY648846.1 | - | 6 - 7 | 839 | 84 | ||
| BC046899.1 | - | 6 - 7 | 839 | 84 | ||
| BC057463.1 | - | 6 - 7 | 820 | 84 | ||
More Information
Set Membership: |
|

mars1
Genomic Map