Assay Details
| Gene Symbol: calm1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200082.1 | NP_956376.1 | 5 - 6 | 485 | 106 | |
| GenBank mRNA | BC059500.1 | - | 5 - 6 | 485 | 106 | |
| BC065426.1 | - | 5 - 6 | 479 | 106 | ||
More Information
Set Membership: |
|

calm1b
Genomic Map