Assay Details
| Gene Symbol: aph1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200115.1 | NP_956409.1 | 3 - 4 | 588 | 68 | |
| GenBank mRNA | AF512428.1 | - | 3 - 4 | 588 | 68 | |
| AY423036.1 | - | 3 - 4 | 579 | 68 | ||
| BC071492.1 | - | 3 - 4 | 582 | 68 | ||
More Information
Set Membership: |
|

aph1b
Genomic Map