Assay Details
| Gene Symbol: scn1laa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200132.2 | NP_956426.2 | 19 - 20 | 3723 | 56 | |
| XM_017357661.2 | XP_017213150.1 | 20 - 21 | 4062 | 56 | ||
| GenBank mRNA | DQ149503.1 | - | 19 - 20 | 3723 | 56 | |
More Information
Set Membership: |
|

scn1laa
Genomic Map