Assay Details
| Gene Symbol: rasl11b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200140.1 | NP_956434.1 | 3 - 4 | 432 | 99 | |
| GenBank mRNA | BC044471.1 | - | 3 - 4 | 432 | 99 | |
| BC065446.1 | - | 3 - 4 | 403 | 99 | ||
| DQ983377.1 | - | 3 - 4 | 416 | 99 | ||
More Information
Set Membership: |
|

rasl11b
Genomic Map