Assay Details
| Gene Symbol: rnaseh2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200226.2 | NP_956520.2 | 6 - 7 | 769 | 55 | |
| GenBank mRNA | AY391457.1 | - | 5 - 6 | 568 | 55 | |
| BC046061.1 | - | 6 - 7 | 595 | 55 | ||
| BC071375.1 | - | 6 - 7 | 769 | 55 | ||
More Information
Set Membership: |
|

rnaseh2a
Genomic Map