Assay Details
| Gene Symbol: polm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200248.1 | NP_956542.1 | 3 - 4 | 523 | 65 | |
| GenBank mRNA | AY817757.1 | - | 3 - 4 | 65 | ||
| BC047832.1 | - | 3 - 4 | 523 | 65 | ||
| BC165363.1 | - | 3 - 4 | 526 | 65 | ||
More Information
Set Membership: |
|

polm
Genomic Map