Assay Details
| Gene Symbol: st3gal3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200355.1 | NP_956649.1 | 3 - 4 | 311 | 86 | |
| XM_021476802.1 | XP_021332477.1 | 3 - 4 | 404 | 86 | ||
| XM_021476804.1 | XP_021332479.1 | 3 - 4 | 634 | 86 | ||
| GenBank mRNA | AJ626820.1 | - | 3 - 4 | 338 | 86 | |
| AJ705063.1 | - | 2 - 3 | 159 | 86 | ||
| BC053179.1 | - | 3 - 4 | 311 | 86 | ||
More Information
Set Membership: |
|

st3gal3a
Genomic Map