Assay Details
| Gene Symbol: st3gal3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200355.1 | NP_956649.1 | 8 - 9 | 696 | 67 | |
| XM_021476802.1 | XP_021332477.1 | 8 - 9 | 789 | 67 | ||
| XM_021476804.1 | XP_021332479.1 | 8 - 9 | 1019 | 67 | ||
| XM_021476805.1 | XP_021332480.1 | 7 - 8 | 707 | 67 | ||
| XM_021476806.1 | XP_021332481.1 | 7 - 8 | 855 | 67 | ||
| XM_021476807.1 | XP_021332482.1 | 9 - 10 | 843 | 67 | ||
| GenBank mRNA | AJ626820.1 | - | 8 - 9 | 723 | 67 | |
| AJ705063.1 | - | 7 - 8 | 544 | 67 | ||
| BC053179.1 | - | 8 - 9 | 696 | 67 | ||
More Information
Set Membership: |
|

st3gal3a
Genomic Map