Assay Details
| Gene Symbol: st3gal3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200355.1 | NP_956649.1 | 9 - 10 | 888 | 87 | |
| GenBank mRNA | AJ626820.1 | - | 9 - 10 | 915 | 87 | |
| AJ705063.1 | - | 8 - 9 | 736 | 87 | ||
| BC053179.1 | - | 9 - 10 | 888 | 87 | ||
More Information
Set Membership: |
|

st3gal3a
Genomic Map