Assay Details
| Gene Symbol: slc35d1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200475.2 | NP_956769.2 | 11 - 12 | 1029 | 138 | |
| GenBank mRNA | BC055186.1 | - | 11 - 12 | 1029 | 137 | |
More Information
Set Membership: |
|

slc35d1b
Genomic Map