Assay Details
| Gene Symbol: parp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200501.1 | NP_956795.1 | 7 - 8 | 1086 | 78 | |
| GenBank mRNA | BC045935.1 | - | 7 - 8 | 1050 | 78 | |
| BC055518.1 | - | 7 - 8 | 1086 | 78 | ||
More Information
Set Membership: |
|

parp3
Genomic Map