Assay Details
| Gene Symbol: myoz2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200899.1 | NP_957193.1 | 4 - 5 | 384 | 84 | |
| XM_017356832.2 | XP_017212321.1 | 5 - 6 | 422 | 84 | ||
| GenBank mRNA | BC064307.1 | - | 4 - 5 | 384 | 84 | |
More Information
Set Membership: |
|

myoz2a
Genomic Map