Assay Details
| Gene Symbol: myoz2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200899.1 | NP_957193.1 | 5 - 6 | 578 | 106 | |
| GenBank mRNA | BC064307.1 | - | 5 - 6 | 578 | 106 | |
More Information
Set Membership: |
|

myoz2a
Genomic Map