Assay Details
| Gene Symbol: ube2e3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_200921.3 | NP_957215.2 | 4 - 5 | 735 | 84 | |
| GenBank mRNA | BC042331.1 | - | 4 - 5 | 497 | 84 | |
| BC067146.1 | - | 4 - 5 | 487 | 84 | ||
More Information
Set Membership: |
|

ube2e3
Genomic Map