Assay Details
| Gene Symbol: epc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201075.1 | NP_957369.1 | 8 - 9 | 1440 | 59 | |
| GenBank mRNA | BC048890.1 | - | 8 - 9 | 1440 | 59 | |
More Information
Set Membership: |
|

epc2
Genomic Map