Assay Details
| Gene Symbol: lin28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201091.1 | NP_957385.1 | 2 - 3 | 367 | 59 | |
| XM_005159535.4 | XP_005159592.1 | 2 - 3 | 471 | 59 | ||
| XM_009294134.3 | XP_009292409.1 | 2 - 3 | 425 | 59 | ||
| GenBank mRNA | BC044433.1 | - | 2 - 3 | 367 | 59 | |
More Information
Set Membership: |
|

lin28a
Genomic Map