Assay Details
| Gene Symbol: sh3gl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201116.1 | NP_957410.1 | 6 - 7 | 724 | 73 | |
| GenBank mRNA | BC044199.1 | - | 6 - 7 | 724 | 73 | |
| BC164387.1 | - | 6 - 7 | 656 | 73 | ||
More Information
Set Membership: |
|

sh3gl2a
Genomic Map