Assay Details
| Gene Symbol: rab1ba | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201142.1 | NP_957436.1 | 3 - 4 | 461 | 110 | |
| GenBank mRNA | BC047816.1 | - | 3 - 4 | 461 | 110 | |
| BC165006.1 | - | 3 - 4 | 223 | 110 | ||
More Information
Set Membership: |
|

rab1ba
Genomic Map