Assay Details
| Gene Symbol: rab32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201332.1 | NP_958489.1 | 2 - 3 | 579 | 71 | |
| GenBank mRNA | BC049531.1 | - | 2 - 3 | 579 | 71 | |
| BC066502.1 | - | 2 - 3 | 567 | 71 | ||
More Information
Set Membership: |
|

rab32a
Genomic Map